Assay Details
Target Gene Details
Entrez Gene ID: | 26512 |
Gene Name: | integrator complex subunit 6 |
Gene Aliases: |
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2 |
Location: |
Chr.13:51353171-51453236 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS6 | NM_001039937.1 | NP_001035026.1 | ||
| NM_001306091.1 | NP_001293020.1 | |||
| NM_012141.2 | NP_036273.1 | |||
| XM_011535040.2 | XP_011533342.1 | |||
| XM_011535041.1 | XP_011533343.1 | |||
| XM_017020540.1 | XP_016876029.1 | |||
| AF097645.1 | AAF03046.1 | |||
| AF141326.1 | AAD39481.1 | |||
| AK074946.1 | ||||
| AK096696.1 | ||||
| AK128795.1 | ||||
| AK296801.1 | ||||
| AK304218.1 | ||||
| AL117626.1 | CAB56020.1 | |||
| AL833524.1 | ||||
| BC013358.2 | AAH13358.1 | |||
| BC039829.1 | AAH39829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map