Assay Details
Target Gene Details
Entrez Gene ID: | 9870 |
Gene Name: | apoptosis resistant E3 ubiquitin protein ligase 1 |
Gene Aliases: |
FIEL1, KIAA0317 |
Location: |
Chr.14:74661252-74713118 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AREL1 | NM_001039479.1 | 20 | 5223 | NP_001034568.1 |
| AB002315.1 | 20 | 5189 | ||
| AK296912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832831 | Chr.14:74546343 - 74740664 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map