Assay Details
Target Gene Details
Entrez Gene ID: | 115426 |
Gene Name: | ubiquitin like with PHD and ring finger domains 2 |
Gene Aliases: |
NIRF, RNF107, TDRD23, URF2 |
Location: |
Chr.9:6413148-6508108 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF2 | NM_152896.2 | NP_690856.1 | ||
| NR_046386.1 | ||||
| XM_011517704.2 | XP_011516006.1 | |||
| XM_011517705.2 | XP_011516007.1 | |||
| XM_017014253.1 | XP_016869742.1 | |||
| XM_017014254.1 | XP_016869743.1 | |||
| AB071698.1 | BAB68317.1 | |||
| AF274049.1 | AAM33799.1 | |||
| AK308433.1 | ||||
| BC028397.2 | AAH28397.1 | |||
| BG709183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831497 | Chr.9:6377501 - 6612402 on Build GRCh38 | Gain |
|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
| dgv7386n100 | Chr.9:6188652 - 6687205 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map