Assay Details
Target Gene Details
Entrez Gene ID: | 26297 |
Gene Name: | secretion regulating guanine nucleotide exchange factor |
Gene Aliases: |
DELGEF, Gnefr |
Location: |
Chr.11:17788048-18013162 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERGEF | NM_012139.3 | 11 | 1495 | NP_036271.1 |
| NR_104040.1 | 12 | 1580 | ||
| NR_104041.1 | 10 | 1328 | ||
| AF086056.1 | 2 | 320 | ||
| AJ243950.1 | 11 | 1361 | CAB60832.1 | |
| AJ243951.1 | 10 | 1194 | CAB60833.1 | |
| AK292286.1 | 11 | 1495 | ||
| BC065375.1 | 11 | 1475 | AAH65375.1 | |
| CB051322.1 | 1 | 103 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553589 | Chr.11:17723445 - 17788177 on Build GRCh38 | Loss |
|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| nsv509390 | Chr.11:17726193 - 17855296 on Build GRCh38 | Insertion |
|
| nsv832078 | Chr.11:17758956 - 17931974 on Build GRCh38 | Loss |
|
| nsv516767 | Chr.11:17737040 - 17788177 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map