Assay Details
Target Gene Details
Entrez Gene ID: | 25959 |
Gene Name: | KN motif and ankyrin repeat domains 2 |
Gene Aliases: |
ANKRD25, MXRA3, PPKWH, SIP |
Location: |
Chr.19:11164267-11197567 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK2 | NM_001136191.2 | 7 | 1939 | NP_001129663.1 |
| NM_015493.6 | 5 | 1681 | NP_056308.3 | |
| AB040951.2 | 6 | 1891 | ||
| AB284125.1 | 8 | 2122 | ||
| AK002094.1 | 4 | 371 | BAA92081.1 | |
| AK023332.1 | 4 | 423 | BAB14531.1 | |
| AY639929.1 | 5 | 1602 | AAT57879.1 | |
| BC082762.1 | 7 | 1893 | AAH82762.1 | |
| BC098105.1 | 6 | 1691 | AAH98105.1 | |
| BC098286.1 | 6 | 1691 | AAH98286.1 | |
| BC098312.1 | 6 | 1691 | AAH98312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065030 | Chr.19:11149372 - 11199804 on Build GRCh38 | Gain |
|
| nsv578613 | Chr.19:11173352 - 11216950 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map