Assay Details
Target Gene Details
Entrez Gene ID: | 8942 |
Gene Name: | kynureninase |
Gene Aliases: |
KYNUU |
Location: |
Chr.2:142877626-143042316 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYNU | NM_001032998.1 | NP_001028170.1 | ||
| NM_001199241.1 | NP_001186170.1 | |||
| NM_003937.2 | NP_003928.1 | |||
| XM_017005217.1 | XP_016860706.1 | |||
| AK223426.1 | BAD97146.1 | |||
| AK290900.1 | ||||
| AK292095.1 | ||||
| AK315343.1 | ||||
| AL548658.3 | ||||
| BC000879.1 | AAH00879.1 | |||
| CB151988.1 | ||||
| CR457423.1 | CAG33704.1 | |||
| U57721.1 | AAC50650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834395 | Chr.2:142777629 - 142971815 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map