Assay Details
Target Gene Details
Entrez Gene ID: | 92105 |
Gene Name: | integrator complex subunit 4 |
Gene Aliases: |
INT4, MST093 |
Location: |
Chr.11:77874491-77994849 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS4 | NM_033547.3 | NP_291025.3 | ||
| XM_011545352.2 | XP_011543654.1 | |||
| XM_011545353.2 | XP_011543655.1 | |||
| XM_017018560.1 | XP_016874049.1 | |||
| AK000455.1 | ||||
| AK290866.1 | ||||
| AL834171.1 | ||||
| BC009995.2 | AAH09995.1 | |||
| BC015664.2 | AAH15664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv526268 | Chr.11:77977728 - 77987478 on Build GRCh38 | Gain |
|
| nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
|
| nsv8841 | Chr.11:77929998 - 77981532 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map