Assay Details
Target Gene Details
Entrez Gene ID: | 11079 |
Gene Name: | retention in endoplasmic reticulum sorting receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.1:2391314-2405444 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RER1 | NM_007033.4 | NP_008964.3 | ||
| XM_005244713.4 | XP_005244770.2 | |||
| XM_006710306.2 | XP_006710369.1 | |||
| XM_011540542.1 | XP_011538844.1 | |||
| XM_011540543.1 | XP_011538845.1 | |||
| XM_017000131.1 | XP_016855620.1 | |||
| XM_017000132.1 | XP_016855621.1 | |||
| XM_017000133.1 | XP_016855622.1 | |||
| XM_017000134.1 | XP_016855623.1 | |||
| XM_017000135.1 | XP_016855624.1 | |||
| AF091071.1 | AAC72940.1 | |||
| AF157324.1 | AAF67490.1 | |||
| AJ001421.1 | CAA04754.1 | |||
| AK095750.1 | ||||
| AK289658.1 | ||||
| BC004965.1 | AAH04965.1 | |||
| BT007224.1 | AAP35888.1 | |||
| CR456806.1 | CAG33087.1 | |||
| DA409924.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951557 | Chr.1:2266062 - 2564661 on Build GRCh38 | Deletion |
|
| nsv1003233 | Chr.1:2348984 - 2404066 on Build GRCh38 | Gain |
|
| nsv823697 | Chr.1:2389693 - 2405919 on Build GRCh38 | Gain |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
| esv2631918 | Chr.1:2400813 - 2401808 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map