Assay Details
Target Gene Details
Entrez Gene ID: | 79442 |
Gene Name: | leucine rich repeat containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:46515385-46580099 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC2 | XM_011534110.2 | XP_011532412.1 | ||
| BC029118.1 | AAH29118.1 |
Target Gene Details
Entrez Gene ID: | 6997 |
Gene Name: | teratocarcinoma-derived growth factor 1 |
Gene Aliases: |
CR, CRGF, CRIPTO |
Location: |
Chr.3:46574555-46582463 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDGF1 | NM_001174136.1 | NP_001167607.1 | ||
| NM_003212.3 | NP_003203.1 | |||
| AK313319.1 | ||||
| BC022393.1 | AAH22393.1 | |||
| BC067844.1 | AAH67844.1 | |||
| CX755454.1 | ||||
| CX781482.1 | ||||
| CX785985.1 | ||||
| X14253.1 | CAA32467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013306 | Chr.3:46427523 - 46657048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map