Assay Details
Target Gene Details
Entrez Gene ID: | 127933 |
Gene Name: | U2AF homology motif (UHM) kinase 1 |
Gene Aliases: |
KIS, KIST, P-CIP2 |
Location: |
Chr.1:162497174-162529629 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHMK1 | NM_001184763.1 | NP_001171692.1 | ||
| NM_144624.2 | NP_653225.2 | |||
| NM_175866.4 | NP_787062.1 | |||
| AJ536197.1 | CAD60192.1 | |||
| AK058195.1 | ||||
| AK292369.1 | ||||
| AL834136.1 | CAI46199.1 | |||
| BC014917.1 | AAH14917.1 | |||
| BC026046.1 | AAH26046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758973 | Chr.1:162094678 - 162533566 on Build GRCh38 | Gain |
|
| nsv516482 | Chr.1:162138329 - 162905405 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map