Assay Details
Target Gene Details
Entrez Gene ID: | 4595 |
Gene Name: | mutY DNA glycosylase |
Gene Aliases: |
MYH |
Location: |
Chr.1:45329242-45340925 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUTYH | NM_001048171.1 | 1 | 51 | NP_001041636.1 |
| NM_001128425.1 | 1 | 51 | NP_001121897.1 | |
| NM_001293190.1 | 1 | 51 | NP_001280119.1 | |
| NM_001293192.1 | 1 | 51 | NP_001280121.1 | |
| NM_012222.2 | 1 | 51 | NP_036354.1 | |
| XM_017001336.1 | XP_016856825.1 | |||
| XM_017001338.1 | 1 | 506 | XP_016856827.1 | |
| XM_017001339.1 | XP_016856828.1 | |||
| AB032920.1 | BAA89336.1 | |||
| AB032921.1 | BAA89337.1 | |||
| AB032922.1 | BAA89338.1 | |||
| AB032923.1 | BAA89339.1 | |||
| AK293830.1 | ||||
| BC003178.1 | AAH03178.1 | |||
| BM926983.1 | 1 | 51 |
Target Gene Details
Entrez Gene ID: | 114034 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
hCaf1z |
Location: |
Chr.1:45339457-45343978 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOE1 | NM_025077.3 | NP_079353.3 | ||
| XM_005270412.3 | XP_005270469.1 | |||
| XM_005270413.4 | 1 | 773 | XP_005270470.1 | |
| XM_011540569.2 | XP_011538871.1 | |||
| AK024011.1 | BAB14774.1 | |||
| AK093320.1 | 1 | 200 | ||
| AK293704.1 | ||||
| AK298162.1 | ||||
| AY169960.1 | AAN75441.1 | |||
| BC009364.2 | AAH09364.1 | |||
| CR457320.1 | CAG33601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map