Assay Details
Target Gene Details
Entrez Gene ID: | 9179 |
Gene Name: | adaptor related protein complex 4 mu 1 subunit |
Gene Aliases: |
CPSQ3, MU-4, MU-ARP2, SPG50 |
Location: |
Chr.7:100101413-100107180 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4M1 | NM_004722.3 | NP_004713.2 | ||
| XM_005250689.4 | XP_005250746.1 | |||
| XM_005250690.4 | XP_005250747.1 | |||
| XM_006716175.3 | XP_006716238.1 | |||
| XM_011516685.2 | XP_011514987.1 | |||
| XM_017012790.1 | XP_016868279.1 | |||
| XM_017012791.1 | XP_016868280.1 | |||
| AF020796.1 | AAD25869.1 | |||
| AF155158.1 | AAD43328.1 | |||
| AK055686.1 | ||||
| AK293590.1 | ||||
| AK296651.1 | ||||
| AK304489.1 | ||||
| AK315782.1 | ||||
| BC018705.1 | AAH18705.1 | |||
| BX640759.1 | CAE45865.1 | |||
| DC413622.1 | ||||
| Y08387.1 | CAA69667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| nsv951384 | Chr.7:100100678 - 100114877 on Build GRCh38 | Deletion |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74961284] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map