Assay Details
Target Gene Details
Entrez Gene ID: | 80831 |
Gene Name: | apolipoprotein L5 |
Gene Aliases: |
APOL-V, APOLV |
Location: |
Chr.22:35708424-35730279 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL5 | NM_030642.1 | 3 | 368 | NP_085145.1 |
| XM_006724321.3 | 4 | 910 | XP_006724384.1 | |
| XM_017028945.1 | 3 | 786 | XP_016884434.1 | |
| AY014878.1 | 3 | 368 | AAK07723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3612 | Chr.22:35713114 - 35761709 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map