Assay Details
Target Gene Details
Entrez Gene ID: | 1053 |
Gene Name: | CCAAT/enhancer binding protein epsilon |
Gene Aliases: |
C/EBP-epsilon, CRP1 |
Location: |
Chr.14:23117306-23119611 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPE | NM_001805.3 | 2 | 1371 | NP_001796.2 |
| BC035797.1 | 2 | 954 | AAH35797.2 | |
| U48866.1 | 2 | 790 | AAC50709.1 |
More Information
Set Membership: |
|

Genomic Map