Assay Details
Target Gene Details
Entrez Gene ID: | 79084 |
Gene Name: | WD repeat domain 77 |
Gene Aliases: |
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50 |
Location: |
Chr.1:111439890-111449308 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR77 | NM_001317062.1 | 10 | 2407 | NP_001303991.1 |
| NM_001317063.1 | 10 | 2365 | NP_001303992.1 | |
| NM_001317064.1 | 8 | 2179 | NP_001303993.1 | |
| NM_024102.3 | 10 | 2371 | NP_077007.1 | |
| NR_133654.1 | 9 | 2229 | ||
| BC016946.1 | 10 | 2271 | AAH16946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819848 | Chr.1:111432982 - 111440610 on Build GRCh38 | Loss |
|
| nsv547535 | Chr.1:111195294 - 111519590 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map