Assay Details
Target Gene Details
Entrez Gene ID: | 340061 |
Gene Name: | transmembrane protein 173 |
Gene Aliases: |
ERIS, MITA, MPYS, NET23, SAVI, STING, hMITA, hSTING |
Location: |
Chr.5:139475528-139482790 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM173 | NM_001301738.1 | 2 | 178 | NP_001288667.1 |
| NM_198282.3 | 3 | 419 | NP_938023.1 | |
| XM_005268445.3 | 3 | 349 | XP_005268502.1 | |
| XM_011537639.2 | 3 | 349 | XP_011535941.1 | |
| XM_011537640.2 | XP_011535942.1 | |||
| AK095896.1 | 3 | 327 | ||
| AK290661.1 | 3 | 356 | ||
| AX775913.1 | 3 | 327 | CAE11656.1 | |
| BC047779.1 | 3 | 336 | AAH47779.1 | |
| BE548954.1 | 3 | 417 | ||
| BM149879.1 | ||||
| FJ222241.1 | 1 | 85 | ||
| KF430638.1 | 1 | 85 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519808 | Chr.5:139396890 - 139805924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map