Assay Details
Target Gene Details
Entrez Gene ID: | 1021 |
Gene Name: | cyclin dependent kinase 6 |
Gene Aliases: |
MCPH12, PLSTIRE |
Location: |
Chr.7:92604921-92836627 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK6 | NM_001145306.1 | 8 | 10192 | NP_001138778.1 |
| NM_001259.6 | 8 | 10087 | NP_001250.1 | |
| XM_006715835.2 | 8 | 10312 | XP_006715898.1 | |
| AA312213.1 | 1 | 483 |
More Information
Set Membership: |
|

Genomic Map