Assay Details
Target Gene Details
Entrez Gene ID: | 81542 |
Gene Name: | thioredoxin related transmembrane protein 1 |
Gene Aliases: |
PDIA11, TMX, TXNDC, TXNDC1 |
Location: |
Chr.14:51240168-51257655 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMX1 | NM_030755.4 | NP_110382.3 | ||
| AB048246.1 | BAB20629.1 | |||
| AK075395.1 | BAC11593.1 | |||
| AK302416.1 | ||||
| AK303134.1 | ||||
| AK312905.1 | ||||
| AL080080.1 | CAB45700.2 | |||
| AY358640.1 | AAQ89003.1 | |||
| BC036460.2 | AAH36460.1 | |||
| BC056874.1 | ||||
| DA203669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952838 | Chr.14:51239683 - 51250782 on Build GRCh38 | Duplication |
|
| esv3584426 | Chr.14:51188530 - 51322710 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map