Assay Details
Target Gene Details
Entrez Gene ID: | 4901 |
Gene Name: | neural retina leucine zipper |
Gene Aliases: |
D14S46E, NRL-MAF, RP27 |
Location: |
Chr.14:24078693-24114924 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRL | NM_006177.3 | 2 | 197 | NP_006168.1 |
| XM_005267708.4 | 3 | 522 | XP_005267765.1 | |
| XM_005267709.3 | 3 | 400 | XP_005267766.1 | |
| XM_005267710.3 | 2 | 174 | XP_005267767.1 | |
| XM_011536801.2 | 2 | 385 | XP_011535103.2 | |
| XM_011536802.1 | 3 | 1532 | XP_011535104.1 | |
| XM_011536804.2 | 3 | 604 | XP_011535106.1 | |
| XM_011536805.2 | 2 | 239 | XP_011535107.1 | |
| XM_011536806.2 | XP_011535108.2 | |||
| XM_017021350.1 | XP_016876839.1 | |||
| AB593102.1 | 2 | 210 | ||
| AB593104.1 | 2 | 208 | ||
| AB593106.1 | 2 | 208 | ||
| BC012395.1 | 2 | 199 | AAH12395.1 | |
| BT006942.1 | 1 | 66 | AAP35588.1 | |
| BX161381.1 | 2 | 385 | CAD61873.1 | |
| BX161522.1 | 3 | 510 | CAD61954.1 | |
| M81840.1 | 2 | 197 | AAA59948.1 | |
| M95925.1 | 2 | 183 | AAA96828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
| nsv1036658 | Chr.14:24044696 - 24086501 on Build GRCh38 | Loss |
|
| nsv1052180 | Chr.14:23960089 - 24086501 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map