Assay Details
Target Gene Details
Entrez Gene ID: | 79000 |
Gene Name: | aurora kinase A and ninein interacting protein |
Gene Aliases: |
AIBP, C1orf135 |
Location: |
Chr.1:25831913-25859458 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AUNIP | NM_001287490.1 | NP_001274419.1 | ||
| NM_024037.2 | 3 | 1967 | NP_076942.1 | |
| XM_017002300.1 | 3 | 2038 | XP_016857789.1 | |
| AI198661.1 | 1 | 212 | ||
| AK024326.1 | 3 | 1866 | BAB14886.1 | |
| AK223419.1 | 3 | 1903 | BAD97139.1 | |
| GQ844687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7490 | Chr.1:25797857 - 25843537 on Build GRCh38 | Deletion |
|
| nsv545883 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147878262] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map