Assay Details
Target Gene Details
Entrez Gene ID: | 9682 |
Gene Name: | lysine demethylase 4A |
Gene Aliases: |
JHDM3A, JMJD2, JMJD2A, TDRD14A |
Location: |
Chr.1:43650118-43705518 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4A | NM_014663.2 | NP_055478.2 | ||
| XM_005271354.3 | XP_005271411.1 | |||
| XM_005271355.3 | XP_005271412.1 | |||
| XM_005271356.3 | XP_005271413.1 | |||
| XM_017002912.1 | XP_016858401.1 | |||
| AB014577.1 | ||||
| AK300039.1 | ||||
| BC002558.2 | AAH02558.1 |
More Information
Set Membership: |
|

Genomic Map