Assay Details
Target Gene Details
Entrez Gene ID: | 25949 |
Gene Name: | SYF2 pre-mRNA splicing factor |
Gene Aliases: |
CBPIN, NTC31, P29, fSAP29 |
Location: |
Chr.1:25222276-25232522 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYF2 | NM_015484.4 | NP_056299.1 | ||
NM_207170.3 | NP_997053.1 | |||
AF273089.1 | AAG42073.1 | |||
AK303582.1 | ||||
AK314859.1 | ||||
AL080166.1 | CAB45754.1 | |||
BC010862.1 | AAH10862.1 | |||
BG390792.1 | ||||
BI668848.1 | ||||
CV806249.1 | ||||
DR155437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv132n100 | Chr.1:24981181 - 25393108 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3576582 | Chr.1:24981195 - 25361084 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1160388 | Chr.1:25177805 - 25266637 on Build GRCh38 | Duplication |
![]() ![]() |
dgv134n100 | Chr.1:25232069 - 25320483 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv133n100 | Chr.1:25228018 - 25361785 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
