Assay Details
Target Gene Details
Entrez Gene ID: | 123920 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 3 |
Gene Aliases: |
BNAS2, CKLFSF3 |
Location: |
Chr.16:66604032-66613892 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM3 | NM_144601.3 | 6 | 1569 | NP_653202.1 |
| NM_181553.2 | 5 | 1405 | NP_853531.1 | |
| NR_037613.1 | 6 | 1789 | ||
| NR_037619.1 | 5 | 1462 | ||
| XM_006721131.1 | 6 | 1613 | XP_006721194.1 | |
| XM_011522861.1 | 4 | 1242 | XP_011521163.1 | |
| AF479813.1 | 6 | 1543 | AAN73435.1 | |
| AK056324.1 | 6 | 1789 | BAB71150.1 | |
| AK075075.1 | 4 | 1102 | ||
| AY166714.1 | 2 | 488 | AAN75573.1 | |
| BC023509.1 | 5 | 1311 | ||
| BC023591.2 | 5 | 1303 | AAH23591.2 | |
| BC036042.1 | 5 | 1365 | AAH36042.2 | |
| BC040227.1 | 5 | 1356 | AAH40227.2 | |
| BC047701.1 | 5 | 1405 | AAH47701.1 |
Target Gene Details
Entrez Gene ID: | 146223 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 4 |
Gene Aliases: |
CKLFSF4 |
Location: |
Chr.16:66598178-66696707 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM4 | XM_017022954.1 | XP_016878443.1 |
More Information
Set Membership: |
|

Genomic Map