Assay Details
Target Gene Details
Entrez Gene ID: | 5069 |
Gene Name: | pappalysin 1 |
Gene Aliases: |
ASBABP2, DIPLA1, IGFBP-4ase, PAPA, PAPP-A, PAPPA1 |
Location: |
Chr.9:116153752-116402321 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPPA | NM_002581.4 | 22 | 6905 | NP_002572.2 |
| XM_006717129.2 | 20 | 4979 | XP_006717192.1 | |
| XM_017014784.1 | 21 | 6831 | XP_016870273.1 | |
| AJ420429.1 | 1 | 1536 | ||
| AK094182.1 | 1 | 666 | ||
| BC078657.1 | 22 | 6535 | AAH78657.1 | |
| BG623020.1 | 1 | 758 | ||
| CB989873.1 |
Target Gene Details
Entrez Gene ID: | 493913 |
Gene Name: | PAPPA antisense RNA 1 |
Gene Aliases: |
DIPAS, NCRNA00156, PAPPA-AS, PAPPAAS, PAPPAS, TAF2C2, TAF4B, TAFII105 |
Location: |
Chr.9:116398157-116400606 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPPA-AS1 | NR_103711.1 | 1 | 2351 | |
| AY623011.1 | 1 | 2351 | AAV41519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7707n100 | Chr.9:116372164 - 116538288 on Build GRCh38 | Gain |
|
| nsv615253 | Chr.9:116368348 - 116409768 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map