Assay Details
Target Gene Details
Entrez Gene ID: | 27173 |
Gene Name: | solute carrier family 39 member 1 |
Gene Aliases: |
ZIP1, ZIRTL |
Location: |
Chr.1:153959099-153968184 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A1 | NM_001271958.1 | NP_001258887.1 | ||
| NM_001271960.1 | NP_001258889.1 | |||
| NM_001271961.1 | NP_001258890.1 | |||
| NM_014437.4 | NP_055252.2 | |||
| AF151829.1 | AAD34066.1 | |||
| AK075257.1 | BAC11502.1 | |||
| BC014303.2 | AAH14303.1 | |||
| DA052424.1 | ||||
| DA818910.1 | ||||
| DB001496.1 |
More Information
Set Membership: |
|

Genomic Map