Assay Details
Target Gene Details
Entrez Gene ID: | 463 |
Gene Name: | zinc finger homeobox 3 |
Gene Aliases: |
ATBF1, ATBT, ZNF927 |
Location: |
Chr.16:72782885-73058635 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFHX3 | NM_001164766.1 | NP_001158238.1 | ||
| NM_006885.3 | NP_008816.3 | |||
| XM_005255957.3 | XP_005256014.1 | |||
| XM_017023251.1 | XP_016878740.1 | |||
| D10250.1 | BAA01095.1 | |||
| L32832.1 | AAC14462.1 |
More Information
Set Membership: |
|

Genomic Map