Assay Details
Target Gene Details
Entrez Gene ID: | 56114 |
Gene Name: | protocadherin gamma subfamily A, 1 |
Gene Aliases: |
PCDH-GAMMA-A1 |
Location: |
Chr.5:141330685-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA1 | NM_018912.2 | NP_061735.1 | ||
| AF152318.1 | AAD43712.1 |
Target Gene Details
Entrez Gene ID: | 56106 |
Gene Name: | protocadherin gamma subfamily A, 10 |
Gene Aliases: |
PCDH-GAMMA-A10 |
Location: |
Chr.5:141413176-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA10 | NM_018913.2 | NP_061736.1 | ||
| AF152319.1 | AAD43713.1 |
Target Gene Details
Entrez Gene ID: | 56105 |
Gene Name: | protocadherin gamma subfamily A, 11 |
Gene Aliases: |
PCDH-GAMMA-A11 |
Location: |
Chr.5:141420970-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA11 | NM_018914.2 | NP_061737.1 | ||
| NM_032092.1 | NP_114481.1 | |||
| AF152320.1 | AAD43714.1 | |||
| AF152504.1 | AAD43765.1 | |||
| BC136781.1 | ||||
| BC171774.1 |
Target Gene Details
Entrez Gene ID: | 26025 |
Gene Name: | protocadherin gamma subfamily A, 12 |
Gene Aliases: |
CDH21, FIB3, PCDH-GAMMA-A12 |
Location: |
Chr.5:141430591-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA12 | NM_003735.2 | NP_003726.1 | ||
| AB011160.1 | ||||
| AF152321.1 | AAD43715.1 |
Target Gene Details
Entrez Gene ID: | 56113 |
Gene Name: | protocadherin gamma subfamily A, 2 |
Gene Aliases: |
PCDH-GAMMA-A2 |
Location: |
Chr.5:141338760-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA2 | NM_018915.3 | NP_061738.1 | ||
| AF152322.1 | AAD43716.1 |
Target Gene Details
Entrez Gene ID: | 56112 |
Gene Name: | protocadherin gamma subfamily A, 3 |
Gene Aliases: |
PCDH-GAMMA-A3 |
Location: |
Chr.5:141344034-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA3 | NM_018916.3 | NP_061739.2 | ||
| AB208927.1 | BAD92164.1 | |||
| AF152323.1 | AAD43717.1 |
Target Gene Details
Entrez Gene ID: | 56111 |
Gene Name: | protocadherin gamma subfamily A, 4 |
Gene Aliases: |
PCDH-GAMMA-A4 |
Location: |
Chr.5:141355025-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA4 | NM_018917.3 | NP_061740.2 | ||
| AF152324.1 | AAD43718.1 |
Target Gene Details
Entrez Gene ID: | 56110 |
Gene Name: | protocadherin gamma subfamily A, 5 |
Gene Aliases: |
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5 |
Location: |
Chr.5:141364331-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA5 | NM_018918.2 | NP_061741.1 | ||
| AF152325.1 | AAD43719.1 |
Target Gene Details
Entrez Gene ID: | 56109 |
Gene Name: | protocadherin gamma subfamily A, 6 |
Gene Aliases: |
PCDH-GAMMA-A6 |
Location: |
Chr.5:141374084-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA6 | NM_018919.2 | NP_061742.1 | ||
| AB209178.1 | BAD92415.1 | |||
| AF152326.1 | AAD43720.1 | |||
| BC146651.1 |
Target Gene Details
Entrez Gene ID: | 56108 |
Gene Name: | protocadherin gamma subfamily A, 7 |
Gene Aliases: |
PCDH-GAMMA-A7 |
Location: |
Chr.5:141382739-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA7 | NM_018920.3 | NP_061743.1 | ||
| AB208958.1 | BAD92195.1 | |||
| AF152327.1 | AAD43721.1 |
Target Gene Details
Entrez Gene ID: | 9708 |
Gene Name: | protocadherin gamma subfamily A, 8 |
Gene Aliases: |
PCDH-GAMMA-A8 |
Location: |
Chr.5:141391916-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA8 | NM_032088.1 | NP_114477.1 | ||
| AF152328.1 | AAD43722.1 |
Target Gene Details
Entrez Gene ID: | 56107 |
Gene Name: | protocadherin gamma subfamily A, 9 |
Gene Aliases: |
PCDH-GAMMA-A9 |
Location: |
Chr.5:141402953-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGA9 | NM_018921.2 | NP_061744.1 | ||
| AF152329.1 | AAD43723.1 |
Target Gene Details
Entrez Gene ID: | 56104 |
Gene Name: | protocadherin gamma subfamily B, 1 |
Gene Aliases: |
PCDH-GAMMA-B1 |
Location: |
Chr.5:141350261-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB1 | NM_018922.2 | NP_061745.1 | ||
| AF152330.1 | AAD43724.1 |
Target Gene Details
Entrez Gene ID: | 56103 |
Gene Name: | protocadherin gamma subfamily B, 2 |
Gene Aliases: |
PCDH-GAMMA-B2 |
Location: |
Chr.5:141360136-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB2 | NM_018923.2 | NP_061746.1 | ||
| AF152331.1 | AAD43725.1 |
Target Gene Details
Entrez Gene ID: | 56102 |
Gene Name: | protocadherin gamma subfamily B, 3 |
Gene Aliases: |
PCDH-GAMMA-B3 |
Location: |
Chr.5:141370264-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB3 | NM_018924.4 | NP_061747.2 | ||
| AF152332.1 | AAD43726.1 | |||
| BC150172.1 |
Target Gene Details
Entrez Gene ID: | 8641 |
Gene Name: | protocadherin gamma subfamily B, 4 |
Gene Aliases: |
CDH20, FIB2, PCDH-GAMMA-B4 |
Location: |
Chr.5:141387885-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB4 | NM_003736.2 | NP_003727.1 | ||
| AF152333.1 | AAD43727.1 |
Target Gene Details
Entrez Gene ID: | 56101 |
Gene Name: | protocadherin gamma subfamily B, 5 |
Gene Aliases: |
PCDH-GAMMA-B5 |
Location: |
Chr.5:141398128-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB5 | NM_018925.2 | NP_061748.1 | ||
| AF152334.1 | AAD43728.1 |
Target Gene Details
Entrez Gene ID: | 56100 |
Gene Name: | protocadherin gamma subfamily B, 6 |
Gene Aliases: |
PCDH-GAMMA-B6 |
Location: |
Chr.5:141408203-141512979 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB6 | NM_018926.2 | NP_061749.1 | ||
| AF152335.1 | AAD43729.1 |
Target Gene Details
Entrez Gene ID: | 56099 |
Gene Name: | protocadherin gamma subfamily B, 7 |
Gene Aliases: |
ME6, PCDH-GAMMA-B7 |
Location: |
Chr.5:141417647-141512981 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGB7 | NM_018927.3 | NP_061750.1 | ||
| AF152336.1 | AAD43730.1 | |||
| BC051788.2 | AAH51788.1 |
Target Gene Details
Entrez Gene ID: | 5098 |
Gene Name: | protocadherin gamma subfamily C, 3 |
Gene Aliases: |
PC43, PCDH-GAMMA-C3, PCDH2 |
Location: |
Chr.5:141476002-141512977 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHGC3 | NM_002588.3 | NP_002579.2 | ||
| NM_032402.1 | NP_115778.1 | |||
| NM_032403.2 | NP_115779.1 | |||
| BC006439.1 | AAH06439.1 | |||
| BC019299.1 | AAH19299.1 | |||
| BC026218.1 | AAH26218.1 | |||
| BC027451.1 | AAH27451.1 | |||
| BC063437.1 | AAH63437.1 | |||
| BG765565.1 | ||||
| L11373.1 | AAA75391.1 |
More Information
Set Membership: |
|

Genomic Map