Assay Details
Target Gene Details
Entrez Gene ID: | 1655 |
Gene Name: | DEAD-box helicase 5 |
Gene Aliases: |
G17P1, HLR1, HUMP68, p68 |
Location: |
Chr.17:64498254-64507038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX5 | NM_001320595.1 | NP_001307524.1 | ||
| NM_001320596.1 | NP_001307525.1 | |||
| NM_001320597.1 | NP_001307526.1 | |||
| NM_004396.4 | NP_004387.1 | |||
| AB209919.1 | BAD93156.1 | |||
| AB451257.1 | ||||
| AB451382.1 | ||||
| AK297192.1 | ||||
| AK297753.1 | ||||
| AK297905.1 | ||||
| AK310318.1 | ||||
| BC016027.1 | AAH16027.1 | |||
| BT006943.1 | AAP35589.1 | |||
| BX571764.1 | CAE11890.1 | |||
| X15729.1 | CAA33751.1 | |||
| X52104.1 | CAA36324.1 |
More Information
Set Membership: |
|

Genomic Map