Assay Details
Target Gene Details
Entrez Gene ID: | 665 |
Gene Name: | BCL2 interacting protein 3 like |
Gene Aliases: |
BNIP3a, NIX |
Location: |
Chr.8:26383007-26413128 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIP3L | NM_004331.2 | NP_004322.1 | ||
| XM_005273617.3 | XP_005273674.1 | |||
| AB004788.1 | BAA28692.1 | |||
| AF060922.1 | AAG43134.1 | |||
| AF067396.1 | AAD03589.1 | |||
| AF079221.1 | AAC27723.1 | |||
| AF452712.2 | AAL50978.1 | |||
| AF536326.1 | AAN04051.1 | |||
| AK290213.1 | ||||
| AK300286.1 | ||||
| AK300727.1 | ||||
| AK307195.1 | ||||
| AK315870.1 | ||||
| AK316315.1 | ||||
| AL132665.1 | CAI46217.1 | |||
| BC001559.1 | AAH01559.1 | |||
| BC009603.1 | AAH09603.1 | |||
| BT019500.1 | AAV38307.1 | |||
| BT019501.1 | AAV38308.1 | |||
| CR456701.1 | CAG32982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831273 | Chr.8:26358989 - 26513275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map