Assay Details
Target Gene Details
Entrez Gene ID: | 51010 |
Gene Name: | exosome component 3 |
Gene Aliases: |
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10 |
Location: |
Chr.9:37779714-37785092 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC3 | NM_001002269.2 | NP_001002269.1 | ||
| NM_016042.3 | NP_057126.2 | |||
| AF281132.1 | AAF82133.1 | |||
| AK289571.1 | ||||
| AK290864.1 | ||||
| BC002437.2 | AAH02437.1 | |||
| BC008880.2 | AAH08880.1 | |||
| BP237222.1 | ||||
| CF994051.1 | ||||
| DA352003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891658 | Chr.9:37689626 - 37790130 on Build GRCh38 | Gain |
|
| nsv831552 | Chr.9:37693652 - 37859980 on Build GRCh38 | Gain |
|
| nsv469814 | Chr.9:37777286 - 37950154 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115431773] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map