Assay Details
Target Gene Details
Entrez Gene ID: | 100287042 |
Gene Name: | uncharacterized LOC100287042 |
Gene Aliases: |
- |
Location: |
Chr.17:75271299-75273895 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287042 | NR_036520.1 | 1 | 1929 |
Target Gene Details
Entrez Gene ID: | 60386 |
Gene Name: | solute carrier family 25 member 19 |
Gene Aliases: |
DNC, MCPHA, MUP1, THMD3, THMD4, TPC |
Location: |
Chr.17:75272980-75289957 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A19 | NM_001126121.1 | 8 | 1411 | NP_001119593.1 |
| NM_001126122.1 | 7 | 1321 | NP_001119594.1 | |
| NM_021734.4 | 8 | 1395 | NP_068380.3 | |
| XM_005257559.3 | 8 | 2033 | XP_005257616.1 | |
| XM_005257560.2 | 8 | 1587 | XP_005257617.1 | |
| XM_005257561.3 | 8 | 1923 | XP_005257618.1 | |
| XM_005257562.2 | 8 | 1570 | XP_005257619.1 | |
| XM_006722007.2 | 8 | 1443 | XP_006722070.1 | |
| XM_011525098.1 | 6 | 1588 | XP_011523400.1 | |
| XM_017024926.1 | 7 | 1985 | XP_016880415.1 | |
| XM_017024927.1 | 6 | 1659 | XP_016880416.1 | |
| XM_017024928.1 | 6 | 1604 | XP_016880417.1 | |
| AA743427.1 | 1 | 256 | ||
| AJ251857.1 | 6 | 1225 | CAC27560.1 | |
| AK075239.1 | 9 | 1417 | BAC11492.1 | |
| AK097882.1 | 6 | 2097 | ||
| AK225881.1 | 6 | 1224 | ||
| AL833387.1 | 8 | 1362 | CAI46136.1 | |
| BC001075.2 | 8 | 1323 | AAH01075.1 | |
| BC005120.1 | 8 | 1323 | AAH05120.1 | |
| BM455504.1 | 5 | 929 | ||
| BU607955.1 | 2 | 265 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map