Assay Details
Target Gene Details
Entrez Gene ID: | 55215 |
Gene Name: | Fanconi anemia complementation group I |
Gene Aliases: |
KIAA1794 |
Location: |
Chr.15:89243954-89317132 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCI | NM_001113378.1 | 17 | 1718 | NP_001106849.1 |
| NM_018193.2 | 17 | 1718 | NP_060663.2 | |
| XM_011521756.2 | 17 | 1742 | XP_011520058.1 | |
| XM_011521757.2 | 17 | 1738 | XP_011520059.1 | |
| XM_011521764.2 | 17 | 1742 | XP_011520066.1 | |
| XM_011521767.2 | 17 | 1712 | XP_011520069.1 | |
| AB058697.1 | 3 | 1642 | BAB47423.1 | |
| AK001581.1 | 11 | 1107 | BAA91770.1 | |
| AK027564.1 | 6 | 605 | BAB55200.1 | |
| AK055176.1 | 17 | 1677 | ||
| BC140769.1 | 17 | 1675 | ||
| BC144483.1 | 17 | 1675 | ||
| EF469766.1 | 16 | 1628 | ||
| EF567077.1 | 16 | 1628 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| nsv833086 | Chr.15:89275748 - 89437378 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map