Assay Details
Target Gene Details
Entrez Gene ID: | 65260 |
Gene Name: | cytochrome c oxidase assembly factor 7 (putative) |
Gene Aliases: |
C1orf163, RESA1, SELRC1 |
Location: |
Chr.1:52686342-52698366 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COA7 | NM_023077.2 | 3 | 1038 | NP_075565.2 |
| AK022501.1 | 3 | 1038 | BAB14063.1 | |
| AK023237.1 | 1 | 323 | ||
| AK225041.1 | 3 | 1013 | ||
| AK225087.1 | 3 | 1019 | ||
| BC015313.1 | AAH15313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| esv2758934 | Chr.1:52660343 - 52823509 on Build GRCh38 | Loss |
|

Genomic Map