Assay Details
Target Gene Details
Entrez Gene ID: | 3621 |
Gene Name: | inhibitor of growth family member 1 |
Gene Aliases: |
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a |
Location: |
Chr.13:110712623-110721074 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ING1 | NM_001267728.1 | 2 | 1371 | NP_001254657.1 |
| NM_005537.5 | 2 | 2276 | NP_005528.4 | |
| NM_198217.2 | 2 | 1634 | NP_937860.1 | |
| NM_198218.2 | 2 | 1603 | NP_937861.1 | |
| NM_198219.2 | 2 | 2302 | NP_937862.1 | |
| AB024401.1 | 2 | 1865 | BAA82886.1 | |
| AB024402.1 | 2 | 2305 | BAA82887.1 | |
| AB031269.1 | 2 | 1720 | BAA83496.1 | |
| AF001954.1 | 2 | 1475 | AAB60879.1 | |
| AF181849.1 | 2 | 2276 | AAF07920.1 | |
| AF181850.1 | 2 | 2287 | AAF07921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map