Assay Details
Target Gene Details
Entrez Gene ID: | 51292 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
GMPR 2 |
Location: |
Chr.14:24232417-24239242 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPR2 | NM_001002000.2 | NP_001002000.1 | ||
| NM_001002001.2 | NP_001002001.1 | |||
| NM_001002002.2 | NP_001002002.1 | |||
| NM_001283021.1 | NP_001269950.1 | |||
| NM_001283022.1 | 1 | 212 | NP_001269951.1 | |
| NM_001283023.1 | NP_001269952.1 | |||
| NM_016576.4 | 1 | 212 | NP_057660.2 | |
| NR_104265.1 | ||||
| XM_005267740.3 | XP_005267797.1 | |||
| XM_005267741.3 | XP_005267798.1 | |||
| XM_005267742.3 | XP_005267799.1 | |||
| XM_006720165.2 | XP_006720228.1 | |||
| XM_017021356.1 | XP_016876845.1 | |||
| XM_017021357.1 | XP_016876846.1 | |||
| XM_017021358.1 | XP_016876847.1 | |||
| XM_017021359.1 | XP_016876848.1 | |||
| AF086919.1 | AAP97149.1 | |||
| AF135159.1 | AAG09132.1 | |||
| AF419346.1 | AAN32701.1 | |||
| AK226166.1 | ||||
| AV660418.1 | ||||
| BC003053.1 | 1 | 161 | AAH03053.1 | |
| BC008021.1 | 1 | 160 | AAH08021.2 | |
| BC009832.2 | AAH09832.1 | |||
| BC093039.1 | AAH93039.1 | |||
| BE003291.1 | ||||
| BI259066.1 | 1 | 174 | ||
| BP347823.1 | ||||
| BX161436.1 | CAD61908.1 | |||
| BX161518.1 | CAD61951.1 | |||
| BX247993.1 | CAD62327.1 | |||
| BX375671.2 | 1 | 183 |
More Information
Set Membership: |
|

Genomic Map