Assay Details
Target Gene Details
Entrez Gene ID: | 51068 |
Gene Name: | NMD3 ribosome export adaptor |
Gene Aliases: |
CGI-07 |
Location: |
Chr.3:161221262-161253532 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMD3 | NM_001320227.1 | NP_001307156.1 | ||
| NM_015938.4 | NP_057022.2 | |||
| XM_005247511.1 | XP_005247568.1 | |||
| XM_005247512.1 | XP_005247569.1 | |||
| AF132941.1 | AAD27716.1 | |||
| AK021775.1 | ||||
| AK094749.1 | ||||
| AK225664.1 | ||||
| AK296710.1 | ||||
| AK308865.1 | ||||
| BC013317.1 | AAH13317.1 | |||
| DB480542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592128 | Chr.3:160795732 - 161829856 on Build GRCh38 | Loss |
|
| nsv1015063 | Chr.3:161200519 - 161306282 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map