Assay Details
Target Gene Details
Entrez Gene ID: | 84893 |
Gene Name: | F-box protein, helicase, 18 |
Gene Aliases: |
FBH1, Fbx18, hFBH1 |
Location: |
Chr.10:5889572-5937595 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO18 | NM_001258452.1 | NP_001245381.1 | ||
| NM_001258453.1 | NP_001245382.1 | |||
| NM_032807.4 | NP_116196.3 | |||
| NM_178150.2 | NP_835363.1 | |||
| XM_011519748.2 | XP_011518050.1 | |||
| XM_017016842.1 | XP_016872331.1 | |||
| XM_017016843.1 | XP_016872332.1 | |||
| XM_017016844.1 | XP_016872333.1 | |||
| XM_017016845.1 | XP_016872334.1 | |||
| AF380349.1 | AAM73631.1 | |||
| AF454502.1 | AAP97700.1 | |||
| AF456237.1 | AAP97705.1 | |||
| AK095343.1 | BAC04535.1 | |||
| AK122753.1 | ||||
| BC110884.1 | AAI10885.1 | |||
| BC113375.1 | ||||
| BC113377.1 | ||||
| DA326341.1 | ||||
| DB099150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831775 | Chr.10:5762372 - 5958886 on Build GRCh38 | Loss |
|
| nsv825229 | Chr.10:5844942 - 5925734 on Build GRCh38 | Gain |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
| nsv549906 | Chr.10:5878873 - 5896581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map