Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC4C | NM_001258419.1 | 7 | 1047 | NP_001245348.1 |
| NM_020929.2 | 5 | 878 | NP_065980.1 | |
| NR_047673.1 | 3 | 1379 | ||
| NR_047674.1 | 2 | 1326 | ||
| XM_011520238.2 | 8 | 1835 | XP_011518540.1 | |
| XM_011520239.2 | 9 | 1901 | XP_011518541.1 | |
| XM_011520240.2 | 7 | 1755 | XP_011518542.1 | |
| XM_011520241.2 | 4 | 2470 | XP_011518543.1 | |
| XM_011520242.2 | 4 | 2456 | XP_011518544.1 | |
| XM_011520243.2 | 7 | 1782 | XP_011518545.1 | |
| XM_011520244.2 | 6 | 1702 | XP_011518546.1 | |
| XM_017018070.1 | 8 | 1831 | XP_016873559.1 | |
| XM_017018071.1 | 8 | 1821 | XP_016873560.1 | |
| XM_017018072.1 | 8 | 1684 | XP_016873561.1 | |
| XM_017018073.1 | 6 | 1543 | XP_016873562.1 | |
| XM_017018074.1 | 4 | 2451 | XP_016873563.1 | |
| XM_017018075.1 | 5 | 1490 | XP_016873564.1 | |
| XM_017018076.1 | 3 | 2417 | XP_016873565.1 | |
| XM_017018077.1 | 6 | 1570 | XP_016873566.1 | |
| XM_017018078.1 | 5 | 2536 | XP_016873567.1 | |
| XM_017018079.1 | 6 | 1698 | XP_016873568.1 | |
| AB046800.1 | 2 | 2311 | BAB13406.1 | |
| AK289646.1 | 3 | 514 | ||
| AY358293.1 | 2 | 1157 | AAQ88660.1 | |
| BC041374.2 | 3 | 1379 | AAH41374.3 | |
| DQ084201.1 | 7 | 996 | AAY86708.1 | |
| DQ084202.1 | 5 | 1015 | AAY86709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map