Assay Details
Target Gene Details
Entrez Gene ID: | 2674 |
Gene Name: | GDNF family receptor alpha 1 |
Gene Aliases: |
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1 |
Location: |
Chr.10:116056925-116273645 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA1 | NM_001145453.2 | 10 | 2872 | NP_001138925.1 |
| NM_005264.5 | 11 | 2887 | NP_005255.1 | |
| NM_145793.3 | 9 | 2421 | NP_665736.1 | |
| XM_005269696.1 | 11 | 2497 | XP_005269753.1 | |
| XM_011539634.1 | 11 | 2462 | XP_011537936.1 | |
| BM675677.1 | 1 | 214 |
More Information
Set Membership: |
|

Genomic Map