Assay Details
Target Gene Details
Entrez Gene ID: | 112703 |
Gene Name: | family with sequence similarity 71 member E1 |
Gene Aliases: |
- |
Location: |
Chr.19:50466643-50476746 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71E1 | NM_001308429.1 | 1 | 428 | NP_001295358.1 |
| NM_138411.2 | 1 | 428 | NP_612420.1 | |
| XM_011526405.2 | 1 | 85 | XP_011524707.2 | |
| XM_011526406.2 | 1 | 85 | XP_011524708.2 | |
| AK125745.1 | ||||
| BC012203.1 | AAH12203.1 | |||
| BC071737.1 | 1 | 428 | AAH71737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|

Genomic Map