Assay Details
Target Gene Details
Entrez Gene ID: | 115948 |
Gene Name: | coiled-coil domain containing 151 |
Gene Aliases: |
CILD30 |
Location: |
Chr.19:11420605-11435782 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC151 | NM_001302453.1 | NP_001289382.1 | ||
| AK302113.1 |
Target Gene Details
Entrez Gene ID: | 5589 |
Gene Name: | protein kinase C substrate 80K-H |
Gene Aliases: |
AGE-R2, G19P1, GIIB, PCLD, PKCSH, PLD1, VASAP-60 |
Location: |
Chr.19:11435257-11450968 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCSH | NM_001001329.2 | 1 | 254 | NP_001001329.1 |
| NM_001289103.1 | 1 | 254 | NP_001276032.1 | |
| NM_001289104.1 | 1 | 254 | NP_001276033.1 | |
| XM_011528130.1 | XP_011526432.1 | |||
| XM_011528131.1 | XP_011526433.1 | |||
| XM_011528132.1 | XP_011526434.1 | |||
| BI254784.1 | 1 | 62 | ||
| DB024838.1 | ||||
| HY029996.1 | ||||
| HY051935.1 | 1 | 174 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144129497] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map