Assay Details
Target Gene Details
Entrez Gene ID: | 5452 |
Gene Name: | POU class 2 homeobox 2 |
Gene Aliases: |
OCT2, OTF2, Oct-2 |
Location: |
Chr.19:42086110-42197931 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU2F2 | NM_001207025.2 | 14 | 2382 | NP_001193954.1 |
| NM_001247994.1 | 14 | 2198 | NP_001234923.1 | |
| NM_002698.4 | 14 | 2334 | NP_002689.1 | |
| XM_005259010.3 | 15 | 2425 | XP_005259067.1 | |
| XM_011527040.2 | 15 | 2539 | XP_011525342.1 | |
| XM_011527041.2 | 14 | 2465 | XP_011525343.1 | |
| XM_011527042.2 | 14 | 2417 | XP_011525344.1 | |
| XM_011527043.2 | 15 | 2626 | XP_011525345.2 | |
| XM_017026884.1 | 16 | 2837 | XP_016882373.1 | |
| XM_017026885.1 | 16 | 2840 | XP_016882374.1 | |
| XM_017026886.1 | 17 | 4339 | XP_016882375.1 | |
| XM_017026887.1 | 17 | 2809 | XP_016882376.1 | |
| XM_017026888.1 | 17 | 3289 | XP_016882377.1 | |
| XM_017026889.1 | 16 | 2831 | XP_016882378.1 | |
| XM_017026890.1 | 16 | 2912 | XP_016882379.1 | |
| XM_017026891.1 | 16 | 2789 | XP_016882380.1 | |
| XM_017026892.1 | 16 | 2771 | XP_016882381.1 | |
| XM_017026893.1 | 15 | 2491 | XP_016882382.1 | |
| XM_017026894.1 | 15 | 2473 | XP_016882383.1 | |
| XM_017026895.1 | 15 | 2471 | XP_016882384.1 | |
| XM_017026896.1 | 15 | 2763 | XP_016882385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map