Assay Details
Target Gene Details
Entrez Gene ID: | 51234 |
Gene Name: | ER membrane protein complex subunit 4 |
Gene Aliases: |
PIG17, TMEM85 |
Location: |
Chr.15:34224997-34230165 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC4 | NM_001286420.1 | 4 | 579 | NP_001273349.1 |
| NM_016454.3 | 5 | 740 | NP_057538.1 | |
| AF151018.1 | 5 | 693 | AAF36104.1 | |
| AK075227.1 | 5 | 719 | BAC11486.1 | |
| AK290524.1 | 4 | 563 | ||
| BC002583.2 | 4 | 544 | AAH02583.1 | |
| BG707804.1 | ||||
| DB506932.1 | 2 | 151 | ||
| KU178640.1 | ||||
| KU178641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain |
|
| esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss |
|
| esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
|
| dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs71702457] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map