Assay Details
Target Gene Details
Entrez Gene ID: | 8611 |
Gene Name: | phospholipid phosphatase 1 |
Gene Aliases: |
LLP1a, LPP1, PAP-2a, PAP2, PPAP2A |
Location: |
Chr.5:55424842-55535370 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP1 | NM_003711.3 | 5 | 1090 | NP_003702.2 |
| NM_176895.2 | 5 | 1093 | NP_795714.1 | |
| NR_103485.1 | 6 | 1245 | ||
| XM_006714724.2 | 4 | 1230 | XP_006714787.1 | |
| AB000888.1 | 5 | 688 | BAA22593.1 | |
| AF014402.1 | 5 | 982 | AAC16032.1 | |
| AF014403.1 | 5 | 985 | AAC16033.1 | |
| AF017116.1 | 5 | 742 | AAC32041.1 | |
| BC039847.1 | 5 | 954 | AAH39847.1 | |
| BC117133.1 | 5 | 691 | ||
| BC143281.1 | 5 | 691 | ||
| BQ019387.1 | 2 | 596 |
More Information
Set Membership: |
|

Genomic Map