Assay Details
Target Gene Details
Entrez Gene ID: | 604 |
Gene Name: | B-cell CLL/lymphoma 6 |
Gene Aliases: |
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51 |
Location: |
Chr.3:187721377-187745725 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL6 | NM_001130845.1 | 5 | 1050 | NP_001124317.1 |
| NM_001134738.1 | 4 | 642 | NP_001128210.1 | |
| NM_001706.4 | 5 | 958 | NP_001697.2 | |
| XM_005247694.3 | 5 | 4302 | XP_005247751.1 | |
| XM_011513062.2 | 5 | 958 | XP_011511364.1 | |
| AK304325.1 | 5 | 701 | ||
| AK308162.1 | 5 | 700 | ||
| BC142705.1 | 5 | 673 | ||
| BC150184.1 | 5 | 674 | ||
| BX649185.1 | 5 | 1579 | CAE46203.1 | |
| EU139066.1 | 3 | 617 | ||
| EU883531.1 | 3 | 593 | ||
| S67779.1 | 5 | 1012 | ||
| U00115.1 | 5 | 920 | AAC50054.1 | |
| Z21943.1 | 5 | 677 | CAA79937.1 |
Target Gene Details
Entrez Gene ID: | 100131635 |
Gene Name: | hCG1645011-like |
Gene Aliases: |
- |
Location: |
Chr.3:187702366-187732415 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100131635 | NR_034062.1 | |||
| AW235368.1 |
More Information
Set Membership: |
|

Genomic Map