Assay Details
Target Gene Details
Entrez Gene ID: | 81832 |
Gene Name: | neuropilin and tolloid like 1 |
Gene Aliases: |
BCTL1, BTCL1 |
Location: |
Chr.18:72742314-72867667 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NETO1 | NM_001201465.1 | NP_001188394.1 | ||
| NM_138966.3 | 11 | 2358 | NP_620416.1 | |
| XM_005266774.4 | 11 | 1877 | XP_005266831.1 | |
| XM_017026020.1 | 11 | 1965 | XP_016881509.1 | |
| XM_017026021.1 | XP_016881510.1 | |||
| AF448838.1 | 11 | 2364 | AAM18026.1 | |
| AL834354.1 | CAD39019.1 | |||
| BC050329.1 | AAH50329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34051 | Chr.18:72637871 - 73006490 on Build GRCh38 | Loss |
|
| nsv577622 | Chr.18:72735518 - 72762852 on Build GRCh38 | Gain |
|
| esv3893139 | Chr.18:70756959 - 72965464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map