Assay Details
Target Gene Details
Entrez Gene ID: | 9122 |
Gene Name: | solute carrier family 16 member 4 |
Gene Aliases: |
MCT4, MCT5 |
Location: |
Chr.1:110362851-110391970 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A4 | NM_001201546.1 | 5 | 817 | NP_001188475.1 |
| NM_001201547.1 | 5 | 828 | NP_001188476.1 | |
| NM_001201548.1 | 4 | 684 | NP_001188477.1 | |
| NM_001201549.1 | NP_001188478.1 | |||
| NM_001319220.1 | NP_001306149.1 | |||
| NM_004696.2 | 6 | 961 | NP_004687.1 | |
| XM_005271317.4 | 7 | 1979 | XP_005271374.1 | |
| XM_006711033.3 | 4 | 1570 | XP_006711096.1 | |
| AK129985.1 | 6 | 905 | ||
| AK223307.1 | 6 | 905 | BAD97027.1 | |
| AK290720.1 | 5 | 761 | ||
| AK295946.1 | 5 | 794 | ||
| AK298539.1 | 4 | 628 | ||
| AK313735.1 | 6 | 923 | ||
| AL833619.1 | 5 | 758 | ||
| BC021664.1 | AAH21664.1 | |||
| BE893159.1 | 5 | 669 | ||
| BX647948.1 | ||||
| U59185.1 | 6 | 893 | AAB72035.1 |
More Information
Set Membership: |
|

Genomic Map