Assay Details
Target Gene Details
Entrez Gene ID: | 353174 |
Gene Name: | zinc activated ion channel |
Gene Aliases: |
L2, LGICZ, LGICZ1, ZAC, ZAC1 |
Location: |
Chr.17:76079182-76082804 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZACN | NM_180990.3 | NP_851321.2 | ||
| AB223030.1 | BAE19924.1 | |||
| AF512521.1 | AAO20969.1 | |||
| AK122638.1 | ||||
| BC110597.1 | AAI10598.1 | |||
| DB119720.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2142 | Chr.17:76070185 - 76087802 on Build GRCh38 | Insertion |
|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
| nsv509674 | Chr.17:76052197 - 76106436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map