Assay Details
Target Gene Details
Entrez Gene ID: | 54988 |
Gene Name: | acyl-CoA synthetase medium-chain family member 5 |
Gene Aliases: |
- |
Location: |
Chr.16:20409508-20441333 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSM5 | NM_001324371.1 | NP_001311300.1 | ||
| NM_001324372.1 | NP_001311301.1 | |||
| NM_001324373.1 | NP_001311302.1 | |||
| NM_017888.2 | NP_060358.2 | |||
| XM_006721056.3 | XP_006721119.1 | |||
| AK000588.1 | BAA91273.1 | |||
| BC013753.1 | AAH13753.1 | |||
| BC016703.1 | AAH16703.1 | |||
| BC068516.1 | AAH68516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047399 | Chr.16:20378305 - 20522307 on Build GRCh38 | Gain |
|
| dgv2795n100 | Chr.16:20325210 - 20411678 on Build GRCh38 | Gain |
|
| nsv984287 | Chr.16:20409387 - 20492193 on Build GRCh38 | Duplication |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| dgv4942n54 | Chr.16:20340615 - 20466152 on Build GRCh38 | Gain |
|
| esv3638138 | Chr.16:20348867 - 20489664 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map