Assay Details
Target Gene Details
Entrez Gene ID: | 7311 |
Gene Name: | ubiquitin A-52 residue ribosomal protein fusion product 1 |
Gene Aliases: |
CEP52, HUBCEP52, L40, RPL40 |
Location: |
Chr.19:18563766-18577460 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA52 | NM_001033930.2 | 1 | 38 | NP_001029102.1 |
| NM_001321017.1 | NP_001307946.1 | |||
| NM_001321018.1 | NP_001307947.1 | |||
| NM_001321019.1 | NP_001307948.1 | |||
| NM_001321020.1 | NP_001307949.1 | |||
| NM_001321022.1 | 1 | 38 | NP_001307951.1 | |
| NM_003333.4 | 1 | 38 | NP_003324.1 | |
| XM_005260052.2 | XP_005260109.1 | |||
| XM_005260053.3 | 1 | 36 | XP_005260110.1 | |
| XM_006722871.2 | XP_006722934.1 | |||
| AF348700.1 | AAK31162.1 | |||
| AK314717.1 | ||||
| BC101830.1 | AAI01831.1 | |||
| BC101832.1 | AAI01833.1 | |||
| BC143668.1 | ||||
| BG035502.1 | ||||
| CB124676.1 | ||||
| DA949569.1 | ||||
| HY153317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833776 | Chr.19:18527218 - 18671454 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map